siRNA Details
Gene Symbol: Ap3s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF084575.1 | - | 4, 5 | 426 | ||
AK002565.1 | - | 4, 5 | 452 | |||
AK137489.1 | - | 4, 5 | 447 | |||
AK152100.1 | - | 4, 5 | 458 | |||
AK184535.1 | - | 2, 3 | 106 | |||
AK194047.1 | - | 2, 3 | 208 | |||
BC012656.1 | - | 4, 5 | 437 | 4187607 | ||
BC108327.1 | - | 4, 5 | 445 | 30713247 | ||
BC131985.1 | - | 4, 5 | 427 | 40130262 | ||
BC145234.1 | - | 4, 5 | 427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
