siRNA Details
Gene Symbol: Atp5f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002960.1 | - | 6 | 602 | ||
AK011312.1 | - | 6 | 616 | |||
AK019459.1 | - | 6 | 619 | |||
AK149176.1 | - | 6 | 588 | |||
AK150730.1 | - | 7 | 800 | |||
AK151250.1 | - | 7 | 800 | |||
AK151574.1 | - | 6 | 593 | |||
AK152253.1 | - | 6 | 619 | |||
AK152829.1 | - | 6 | 619 | |||
AK157544.1 | - | 6 | 656 | |||
AK158569.1 | - | 6 | 643 | |||
AK162311.1 | - | 6 | 585 | |||
AK167480.1 | - | 6 | 622 | |||
AK167626.1 | - | 6 | 593 | |||
AK168028.1 | - | 6 | 618 | |||
AK168041.1 | - | 6 | 619 | |||
AK168590.1 | - | 6 | 593 | |||
AK169137.1 | - | 6 | 620 | |||
AK169634.1 | - | 6 | 575 | |||
BC049640.1 | - | 6 | 550 | |||
BC087882.1 | - | 8 | 1104 | 6706159 | ||
BC089574.1 | - | 6 | 582 | 6756180 |

More Information
Other Species Targeted

