siRNA Details
Gene Symbol: Atp5j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA259963.1 | - | 2 | 160 | ||
AI326720.1 | - | 3 | 427 | |||
AK078484.1 | - | 2 | 321 | |||
BC010766.1 | - | 3 | 456 | 4219715 | ||
BE851144.1 | - | 3 | 198 | |||
BY079469.1 | - | 2 | 235 | |||
U77128.1 | - | 1 | 119 | |||
W75852.1 | - | 2 | 186 |

More Information
Other Species Targeted

