siRNA Details
Gene Symbol: Bcap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164090.1 | NP_001157562.1 | 7 | 737 | ||
NM_007530.3 | NP_031556.1 | 6 | 900 | |||
GenBank mRNA | AK090068.1 | - | 7 | 714 | ||
AK144754.1 | - | 7 | 711 | |||
BC003303.1 | - | 6 | 804 | 3584161 | ||
BC021661.1 | - | 6 | 911 | 2646883 | ||
BC040751.1 | - | 1 | 1090 | |||
X78684.1 | - | 6 | 712 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
