siRNA Details
Gene Symbol: Bcl2a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009742.3 | NP_033872.1 | 1 | 278 | ||
GenBank mRNA | BC028762.1 | - | 224 | 1226745 | ||
CT010250.1 | - | 147 | ||||
L16462.1 | - | 1 | 278 |
Gene Symbol: Bcl2a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007534.3 | NP_031560.2 | 1 | 234 | ||
GenBank mRNA | AK138369.1 | - | 1 | 214 | ||
AK152200.1 | - | 1 | 204 | |||
BC100462.1 | - | 1 | 211 | 30295442 | ||
BC116241.1 | - | 1 | 234 | 40091848 | ||
BE650553.1 | - | 1 | 328 |
Gene Symbol: Bcl2a1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007536.2 | NP_031562.1 | 1 | 278 | ||
GenBank mRNA | AK165576.1 | - | 1 | 30 | ||
BC027536.1 | - | 1 | 191 | 1363928 | ||
BC053965.1 | - | 1 | 300 | |||
BC116240.2 | - | 1 | 234 | 40091847 |

More Information
Other Species Targeted

Gene Ontology Categories:
