siRNA Details
Gene Symbol: Bcl2a1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007536.2 | NP_031562.1 | 1, 2 | 547 | ||
GenBank mRNA | AK165576.1 | - | 1, 2 | 299 | ||
BC027536.1 | - | 1, 2 | 460 | 1363928 | ||
BC053965.1 | - | 1, 2 | 569 | |||
BC116240.2 | - | 1, 2 | 503 | 40091847 |
Gene Symbol: Bcl2a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC028762.1 | - | 493 | 1226745 | ||
CT010250.1 | - | 416 |
Gene Symbol: Bcl2a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007534.3 | NP_031560.2 | 1, 2 | 503 | ||
GenBank mRNA | AK138369.1 | - | 1, 2 | 483 | ||
AK152200.1 | - | 1, 2 | 473 | |||
BC100462.1 | - | 1, 2 | 480 | 30295442 | ||
BC116241.1 | - | 1, 2 | 503 | 40091848 | ||
BP773251.1 | - | 1, 2 | 61 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
