siRNA Details
Gene Symbol: Calca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033954.3 | NP_001029126.1 | 2, 3 | 249 | ||
NM_001289444.1 | NP_001276373.1 | 2, 3 | 273 | |||
NM_007587.2 | NP_031613.2 | 2, 3 | 249 | |||
GenBank mRNA | AF330212.1 | - | 2, 3 | 187 | ||
AK132582.1 | - | 2, 3 | 199 | |||
BC028771.1 | - | 2, 3 | 182 | 1331673 | ||
BC030071.1 | - | 2, 3 | 180 | 1399214 | ||
CJ057527.1 | - | 2, 3 | 224 | |||
CJ060164.1 | - | 2, 3 | 250 | |||
X97991.1 | - | 2, 3 | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
