siRNA Details
Gene Symbol: Calca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033954.3 | NP_001029126.1 | 3, 4 | 381 | ||
NM_001289444.1 | NP_001276373.1 | 3, 4 | 405 | |||
GenBank mRNA | AF330212.1 | - | 3, 4 | 319 | ||
BC028771.1 | - | 3, 4 | 314 | 1331673 | ||
CJ057527.1 | - | 3, 4 | 356 | |||
CJ060164.1 | - | 3, 4 | 382 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
