siRNA Details
Gene Symbol: Cox7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007749.3 | NP_031775.1 | 2 | 279 | ||
GenBank mRNA | AK002494.1 | - | 2 | 246 | ||
AK027989.1 | - | 2 | 241 | |||
AK131871.1 | - | 2 | 275 | |||
AK153444.1 | - | 2 | 251 | |||
AK168077.1 | - | 2 | 267 | |||
AV056265.1 | - | 2 | 119 | |||
BB696274.1 | - | 2 | 289 | |||
BC010772.1 | - | 2 | 261 | 4216583 | ||
BC086792.1 | - | 2 | 251 | 6411273 | ||
X52940.1 | - | 2 | 245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
