siRNA Details
Gene Symbol: Crygd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007776.2 | NP_031802.2 | 3 | 321 | ||
GenBank mRNA | AK136286.1 | - | 3 | 320 | ||
BC057013.1 | - | 3 | 302 | 6824399 | ||
BC132116.1 | - | 3 | 320 | 40130393 | ||
BC132118.1 | - | 3 | 321 | 40130395 | ||
CA452140.1 | - | 2 | 168 | |||
K02583.1 | - | 3 | 290 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
