siRNA Details
Gene Symbol: Celf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244891.1 | NP_001231820.1 | 14 | 1707 | ||
NM_001244903.1 | NP_001231832.1 | 13 | 1654 | |||
NM_017368.3 | NP_059064.2 | 14 | 1806 | |||
NM_198683.2 | NP_941955.1 | 13 | 1654 | |||
GenBank mRNA | AF267535.1 | - | 12 | 1410 | ||
AF314172.1 | - | 12 | 1251 | |||
AJ007987.1 | - | 12 | 1399 | |||
AK014492.1 | - | 13 | 1570 | |||
AK135030.1 | - | 2 | 206 | |||
AK156725.1 | - | 13 | 1567 | |||
AK196827.1 | - | 3 | 352 | |||
AK200025.1 | - | 3 | 362 | |||
AK208664.1 | - | 3 | 348 | |||
BC021393.1 | - | 13 | 1519 | |||
BY719033.1 | - | 2 | 206 | |||
CF745326.1 | - | 3 | 344 |

More Information
Other Species Targeted

Gene Ontology Categories:
