siRNA Details
Gene Symbol: Celf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244891.1 | NP_001231820.1 | 13 | 1562 | ||
NM_001244903.1 | NP_001231832.1 | 12 | 1509 | |||
NM_017368.3 | NP_059064.2 | 13 | 1661 | |||
NM_198683.2 | NP_941955.1 | 12 | 1509 | |||
GenBank mRNA | AF267535.1 | - | 11 | 1265 | ||
AF314172.1 | - | 11 | 1106 | |||
AJ007987.1 | - | 11 | 1254 | |||
AK014492.1 | - | 12 | 1425 | |||
AK135030.1 | - | 1 | 61 | |||
AK143698.1 | - | 13 | 1463 | |||
AK156725.1 | - | 12 | 1422 | |||
AK183548.1 | - | 3 | 269 | |||
AK188588.1 | - | 3 | 268 | |||
AK196827.1 | - | 2 | 207 | |||
AK200025.1 | - | 2 | 217 | |||
AK203078.1 | - | 1 | 82 | |||
AK208664.1 | - | 2 | 203 | |||
BC021393.1 | - | 12 | 1374 | |||
BY719033.1 | - | 1 | 61 | |||
CF745326.1 | - | 2 | 199 | |||
X61451.1 | - | 8 | 959 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
