siRNA Details
Gene Symbol: Edil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037987.3 | NP_001033076.1 | 11 | 1896 | ||
NM_010103.4 | NP_034233.1 | 10 | 1866 | |||
GenBank mRNA | AF031524.1 | - | 11 | 2004 | ||
AK036117.1 | - | 11 | 1857 | |||
AK081003.1 | - | 10 | 1698 | |||
AK139093.1 | - | 11 | 1858 | |||
AK141907.1 | - | 11 | 1880 | |||
BC056386.1 | - | 11 | 1796 | 6854497 |

More Information
Other Species Targeted

Gene Ontology Categories:
