siRNA Details
Gene Symbol: Elk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007923.2 | NP_031949.2 | 3 | 659 | ||
GenBank mRNA | AK054075.1 | - | 3 | 951 | ||
AK156537.1 | - | 3 | 657 | |||
BC144844.1 | - | 3 | 409 | 9053392 | ||
BC145795.1 | - | 3 | 409 | 40131098 | ||
FN434116.1 | - | 2 | 256 | |||
Z36885.1 | - | 4 | 604 |
