siRNA Details
Gene Symbol: Fcgr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077189.1 | NP_001070657.1 | 5 | 721 | ||
NM_010187.2 | NP_034317.1 | 5 | 721 | |||
GenBank mRNA | AK080885.1 | - | 5 | 724 | ||
AK089266.1 | - | 5 | 722 | |||
AK150613.1 | - | 5 | 722 | |||
AK155329.1 | - | 5 | 713 | |||
AK170035.1 | - | 5 | 711 | |||
AK171254.1 | - | 5 | 713 | |||
AK171316.1 | - | 5 | 704 | |||
BC011305.1 | - | 5 | 684 | |||
BC019758.1 | - | 5 | 684 | |||
BC038070.1 | - | 5 | 708 | 3993915 | ||
M14216.1 | - | 5 | 679 | |||
M16367.1 | - | 7 | 962 | |||
M17515.1 | - | 5 | 675 | |||
U31801.1 | - | 3 | 536 | |||
U31802.1 | - | 3 | 536 | |||
U31803.1 | - | 3 | 536 | |||
U31804.1 | - | 3 | 536 | |||
U51629.1 | - | 5 | 629 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
