siRNA Details
Gene Symbol: Fhl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010213.3 | NP_034343.2 | 2 | 271 | ||
GenBank mRNA | AF114382.1 | - | 2 | 242 | ||
AF134772.1 | - | 2 | 268 | |||
AF149826.1 | - | 1 | 69 | |||
AI426098.1 | - | 2 | 268 | |||
BC144903.1 | - | 2 | 235 | 9053451 | ||
BC145939.1 | - | 2 | 235 | 40131242 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
