siRNA Details
Gene Symbol: Flg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076261.1 | - | 2 | 938 | ||
AV230893.2 | - | 1 | 306 | |||
J03458.1 | - | 3 | 1136 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
