siRNA Details
Gene Symbol: Fmo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161763.1 | NP_001155235.1 | 4, 5 | 627 | ||
NM_001161765.1 | NP_001155237.1 | 3, 4 | 710 | |||
NM_010232.4 | NP_034362.2 | 4, 5 | 611 | |||
GenBank mRNA | AK133675.1 | - | 3, 4 | 692 | ||
AK147040.1 | - | 4, 5 | 603 | |||
AK162951.1 | - | 4, 5 | 628 | |||
BC022991.1 | - | 4, 5 | 574 | 5123876 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
