siRNA Details
Gene Symbol: Ftl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010240.2 | NP_034370.2 | 4 | 872 | ||
GenBank mRNA | AK002242.1 | - | 4 | 826 | ||
AK002253.1 | - | 4 | 825 | |||
AK011009.1 | - | 4 | 825 | |||
AK011029.1 | - | 4 | 824 | |||
AK011244.1 | - | 4 | 826 | |||
AK012690.1 | - | 3 | 411 | |||
AK088647.1 | - | 4 | 830 | |||
AK145265.1 | - | 4 | 825 | |||
AK145658.1 | - | 4 | 825 | |||
AK150480.1 | - | 4 | 825 | |||
AK150481.1 | - | 4 | 821 | |||
AK150960.1 | - | 4 | 847 | |||
AK152030.1 | - | 4 | 825 | |||
AK152385.1 | - | 4 | 821 | |||
AK152393.1 | - | 4 | 823 | |||
AK152564.1 | - | 4 | 825 | |||
AK152612.1 | - | 4 | 825 | |||
AK159574.1 | - | 4 | 826 | |||
AK167410.1 | - | 4 | 823 | |||
AK168310.1 | - | 4 | 825 | |||
AK168735.1 | - | 4 | 825 | |||
AK168862.1 | - | 4 | 826 | |||
AK168866.1 | - | 4 | 825 | |||
AK169100.1 | - | 4 | 825 | |||
AK169159.1 | - | 4 | 826 | |||
BC019840.1 | - | 4 | 804 | 5123770 | ||
BC020403.1 | - | 4 | 813 | |||
BC081462.1 | - | 4 | 791 | 6823712 | ||
BC083350.1 | - | 4 | 802 | 6824407 | ||
BC085309.1 | - | 4 | 804 | 6489467 | ||
BC092259.1 | - | 4 | 793 | 5125514 | ||
BC106145.1 | - | 4 | 791 | 5068947 | ||
BC106146.1 | - | 4 | 804 | 6476394 | ||
J04716.1 | - | 4 | 791 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
