siRNA Details
Gene Symbol: G0s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008059.3 | NP_032085.1 | 2 | 621 | ||
GenBank mRNA | AK003165.1 | - | 2 | 543 | ||
BC117038.1 | - | 2 | 449 | 40126357 | ||
BC117040.1 | - | 2 | 449 | 40126359 | ||
X95280.1 | - | 2 | 525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
