siRNA Details
Gene Symbol: Gcsam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159297.1 | NP_001152769.1 | 6 | 488 | ||
NM_008099.4 | NP_032125.3 | 5 | 422 | |||
GenBank mRNA | AK036553.1 | - | 5 | 440 | ||
AY500830.1 | - | 6 | 396 | |||
BC099585.1 | - | 5 | 386 | 30289367 | ||
U13263.1 | - | 5 | 423 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
