siRNA Details
Gene Symbol: Gch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008102.3 | NP_032128.1 | 2 | 496 | ||
GenBank mRNA | AK150657.1 | - | 2 | 508 | ||
AK152571.1 | - | 2 | 508 | |||
BC005643.1 | - | 2 | 496 | 3709238 | ||
BC069921.1 | - | 2 | 500 | 6529016 | ||
L09737.1 | - | 2 | 483 | |||
S71373.1 | - | 2 | 213 |

More Information
Other Species Targeted

Gene Ontology Categories:
