siRNA Details
Gene Symbol: Gsta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008182.3 | NP_032208.2 | 3 | 186 | ||
GenBank mRNA | AK002712.1 | - | 3 | 193 | ||
BC061133.1 | - | 2 | 129 | 30303519 | ||
J03958.1 | - | 3 | 201 |
Gene Symbol: Gm10639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122660.1 | NP_001116132.1 | 3 | 204 | ||
GenBank mRNA | BC030173.1 | - | 3 | 215 | 5041881 |
Gene Symbol: Gm3776 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243092.1 | NP_001230021.1 | 3 | 204 | ||
GenBank mRNA | BC061134.1 | - | 174 | 30308304 |
Gene Symbol: Gsta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008181.3 | NP_032207.3 | 3 | 191 | ||
GenBank mRNA | BC132572.1 | - | 174 | 40130849 | ||
BC132576.1 | - | 174 | 40130853 | |||
BY080629.1 | - | 191 | ||||
CJ228212.1 | - | 1 | 48 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
