siRNA Details
Gene Symbol: H2-T10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010395.7 | NP_034525.4 | 626 | |||
GenBank mRNA | AK153876.1 | - | 615 | |||
AK161731.1 | - | 2 | 931 | |||
BC021751.1 | - | 720 | 4165944 | |||
BC021760.1 | - | 744 | 5055493 | |||
M35244.1 | - | 3 | 730 | |||
M35246.1 | - | 4 | 607 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
