siRNA Details
Gene Symbol: H2-T10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010395.7 | NP_034525.4 | 974 | |||
GenBank mRNA | AK088241.1 | - | 3 | 476 | ||
AK153876.1 | - | 963 | ||||
AK161731.1 | - | 3 | 1290 | |||
BC021751.1 | - | 1068 | 4165944 | |||
BC021760.1 | - | 1092 | 5055493 | |||
M35244.1 | - | 4 | 1078 | |||
M35246.1 | - | 5 | 966 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
