siRNA Details
Gene Symbol: H2-T23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010398.3 | NP_034528.1 | 3 | 563 | ||
GenBank mRNA | AB359230.2 | - | 3 | 563 | ||
AF041855.1 | - | 542 | ||||
AF057279.2 | - | 531 | ||||
AF082510.1 | - | 3 | 531 | |||
AF100695.1 | - | 510 | ||||
AF100696.1 | - | 510 | ||||
AF100699.1 | - | 3 | 504 | |||
AF100700.1 | - | 510 | ||||
AF100701.1 | - | 510 | ||||
AF100702.1 | - | 3 | 510 | |||
AF100703.1 | - | 504 | ||||
AK133502.1 | - | 3 | 563 | |||
BC004093.1 | - | 832 | ||||
BC005648.1 | - | 552 | 3709267 | |||
BC099472.1 | - | 3 | 552 | 30842866 | ||
L00606.1 | - | 557 | ||||
M11284.1 | - | 564 |
Gene Symbol: C920025E04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271005.1 | NP_001257934.1 | 3 | 550 | ||
GenBank mRNA | AK083376.1 | - | 2 | 776 | ||
AK083387.1 | - | 2 | 776 | |||
BG864910.1 | - | 3 | 543 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
