siRNA Details
Gene Symbol: C920025E04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271005.1 | NP_001257934.1 | 3 | 507 | ||
GenBank mRNA | AK083376.1 | - | 2 | 733 | ||
AK083387.1 | - | 2 | 733 | |||
BG864910.1 | - | 3 | 500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
