siRNA Details
Gene Symbol: Hand2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010402.4 | NP_034532.3 | 1 | 1063 | ||
GenBank mRNA | AJ131846.1 | - | 1 | 144 | ||
AK035160.1 | - | 1 | 1065 |

More Information
Other Species Targeted

Gene Ontology Categories:
