siRNA Details
Gene Symbol: Htr1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285482.1 | NP_001272411.1 | 3 | 2153 | ||
NM_001285483.1 | NP_001272412.1 | 3 | 1975 | |||
NM_001285484.1 | NP_001272413.1 | 2 | 1882 | |||
NM_008309.5 | NP_032335.2 | 2 | 1995 | |||
GenBank mRNA | AK082016.1 | - | 2 | 1999 | ||
BC103532.1 | - | 2 | 2006 | 40039729 | ||
BC103535.1 | - | 3 | 1969 | 40039732 | ||
BC103536.1 | - | 3 | 2141 | 40039733 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
