siRNA Details
Gene Symbol: Ido1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293690.1 | NP_001280619.1 | 10 | 1226 | ||
NM_008324.2 | NP_032350.1 | 10 | 1073 | |||
GenBank mRNA | AK033783.1 | - | 10 | 1227 | ||
BC049931.1 | - | 10 | 1064 | 5387937 | ||
M69109.1 | - | 10 | 1130 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
