siRNA Details
Gene Symbol: Cxcl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021274.2 | NP_067249.1 | 2 | 205 | ||
GenBank mRNA | AF227743.1 | - | 2 | 166 | ||
AJ243095.1 | - | 1 | 17 | |||
AK144279.1 | - | 2 | 196 | |||
AK146144.1 | - | 2 | 203 | |||
AK150380.1 | - | 2 | 196 | |||
AK150765.1 | - | 2 | 196 | |||
AK151210.1 | - | 2 | 196 | |||
AK151248.1 | - | 2 | 196 | |||
AK151415.1 | - | 2 | 196 | |||
AK151534.1 | - | 2 | 196 | |||
AK152568.1 | - | 2 | 196 | |||
AK152814.1 | - | 2 | 196 | |||
AK152924.1 | - | 1 | 38 | |||
AK153181.1 | - | 2 | 196 | |||
AK156907.1 | - | 1 | 733 | |||
AK157130.1 | - | 2 | 196 | |||
AK157139.1 | - | 2 | 196 | |||
AK157589.1 | - | 2 | 196 | |||
AK157678.1 | - | 2 | 196 | |||
AK172540.1 | - | 2 | 196 | |||
BC030067.1 | - | 2 | 172 | 1446589 | ||
BC057150.1 | - | 1 | 721 | |||
BY165450.1 | - | 2 | 206 | |||
M33266.1 | - | 2 | 170 | |||
M86829.1 | - | 2 | 195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
