siRNA Details
Gene Symbol: Igfbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008344.3 | NP_032370.2 | 4 | 1252 | ||
GenBank mRNA | AK157982.1 | - | 4 | 936 | ||
BC012723.1 | - | 4 | 959 | 4224052 | ||
X81584.1 | - | 4 | 1082 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
