siRNA Details
Gene Symbol: Il2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008366.3 | NP_032392.1 | 2 | 271 | ||
GenBank mRNA | AF065914.1 | - | 2 | 269 | ||
AF065915.1 | - | 2 | 269 | |||
AF065916.1 | - | 2 | 269 | |||
AY147902.1 | - | 2 | 223 | |||
BC116845.1 | - | 2 | 269 | 40126164 | ||
BC116873.1 | - | 2 | 269 | 40126192 | ||
DQ836354.1 | - | 2 | 163 | |||
K02292.1 | - | 2 | 271 | |||
U41494.1 | - | 2 | 166 | |||
U41504.1 | - | 2 | 181 | |||
U41505.1 | - | 2 | 193 | |||
U41506.1 | - | 2 | 166 | |||
X01772.1 | - | 2 | 270 | |||
X73040.1 | - | 2 | 270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
