siRNA Details
Gene Symbol: Klra7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF288376.1 | - | 6, 7 | 847 | ||
AF419249.1 | - | 1107 | ||||
BC140984.1 | - | 6, 7 | 871 | 9055596 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
