siRNA Details
Gene Symbol: Vmn1r234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134198.1 | NP_598959.1 | 1 | 682 | ||
GenBank mRNA | BC127026.1 | - | 1 | 682 | 40044289 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
