siRNA Details
Gene Symbol: Vmn1r78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134208.2 | NP_598969.2 | 1 | 757 | ||
GenBank mRNA | BC107194.1 | - | 1 | 757 | 40051037 | |
BC107195.1 | - | 1 | 757 | 40051039 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
