siRNA Details
Gene Symbol: Vmn1r222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134240.1 | NP_599001.1 | 1 | 775 | ||
GenBank mRNA | BC120858.1 | - | 1 | 853 | 40129781 | |
BC120860.1 | - | 1 | 853 | 40129783 |
Gene Symbol: Vmn1r188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145850.1 | NP_665849.1 | 1 | 775 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
