siRNA Details
Gene Symbol: Mfge8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045489.1 | NP_001038954.1 | 9 | 1285 | ||
NM_008594.2 | NP_032620.2 | 10 | 1396 | |||
GenBank mRNA | AB021130.2 | - | 10 | 1303 | ||
AB025280.1 | - | 9 | 1192 | |||
AK087555.1 | - | 9 | 1275 | |||
AK089211.1 | - | 9 | 1251 | |||
AK145183.1 | - | 9 | 1251 | |||
AK148371.1 | - | 9 | 1274 | |||
AK152088.1 | - | 10 | 1385 | |||
AK155047.1 | - | 10 | 1394 | |||
AK169997.1 | - | 9 | 1249 | |||
AK170935.1 | - | 9 | 1249 | |||
AK171143.1 | - | 10 | 1360 | |||
AK190819.1 | - | 4 | 416 | |||
BC003892.1 | - | 9 | 1229 | 3593310 | ||
BC003904.1 | - | 9 | 1224 | 3601565 | ||
BC018577.1 | - | 9 | 1222 | |||
BC037475.1 | - | 6 | 1611 | |||
M38337.1 | - | 10 | 1348 | |||
Y11684.2 | - | 9 | 1192 |
