siRNA Details
Gene Symbol: Mif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010798.2 | NP_034928.1 | 3 | 485 | ||
GenBank mRNA | AK008939.1 | - | 3 | 479 | ||
AK010691.1 | - | 3 | 456 | |||
AK013328.1 | - | 3 | 479 | |||
AK152211.1 | - | 3 | 466 | |||
BC024895.1 | - | 3 | 465 | 3978685 | ||
BC086928.1 | - | 3 | 455 | 6762214 | ||
BC100605.1 | - | 3 | 455 | 30118109 | ||
L02913.1 | - | 3 | 458 | |||
L07607.1 | - | 3 | 388 | |||
L10613.1 | - | 3 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
