siRNA Details
Gene Symbol: Mycn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008709.3 | NP_032735.3 | 2 | 343 | ||
GenBank mRNA | AK146696.1 | - | 2 | 344 | ||
AK146913.1 | - | 2 | 341 | |||
BC005453.1 | - | 2 | 227 | 3495446 | ||
BC049783.1 | - | 1 | 272 | 6517457 | ||
M36277.1 | - | 119 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001013096.
Gene Ontology Categories:
