siRNA Details
Gene Symbol: Pla2g1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011107.1 | NP_035237.1 | 4 | 419 | ||
GenBank mRNA | AF094610.1 | - | 4 | 412 | ||
AF097637.1 | - | 4 | 400 | |||
AF162712.1 | - | 4 | 419 | |||
AF187852.1 | - | 4 | 398 | |||
AK007730.1 | - | 4 | 413 | |||
AK007797.1 | - | 4 | 418 | |||
AK008331.1 | - | 4 | 415 | |||
AK008664.1 | - | 4 | 415 | |||
AK008668.1 | - | 4 | 415 | |||
AK008841.1 | - | 4 | 413 | |||
AK008934.1 | - | 4 | 419 | |||
AK008936.1 | - | 4 | 413 | |||
AK009314.1 | - | 4 | 412 | |||
AK028104.1 | - | 4 | 416 | |||
AK028134.1 | - | 4 | 419 | |||
BC145546.1 | - | 3 | 259 | |||
BC145908.1 | - | 4 | 419 | 40131211 | ||
BC145910.1 | - | 4 | 419 | 40131213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
