siRNA Details
Gene Symbol: Pld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164056.1 | NP_001157528.1 | 16 | 1832 | ||
NM_008875.4 | NP_032901.2 | 15 | 1777 | |||
GenBank mRNA | AK136111.1 | - | 10 | 1015 | ||
BC068144.1 | - | 15 | 1634 | 30532470 | ||
U87868.1 | - | 16 | 1815 |
