siRNA Details
Gene Symbol: Ppp3cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008914.2 | NP_032940.1 | 4 | 569 | ||
GenBank mRNA | AB221678.1 | - | 4 | 436 | ||
AK135203.1 | - | 4 | 383 | |||
BC037668.1 | - | 4 | 392 | |||
BC066000.1 | - | 4 | 540 | 5699179 | ||
M81483.1 | - | 4 | 406 |

More Information
Other Species Targeted

Gene Ontology Categories:
