siRNA Details
Gene Symbol: Ppp3cc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133393.1 | - | 14 | 1628 | ||
BC046617.1 | - | 15 | 1460 | |||
BC141079.1 | - | 14 | 1448 | 9055691 | ||
BC145347.1 | - | 13 | 1323 | |||
M81475.1 | - | 14 | 1601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
