siRNA Details
Gene Symbol: Bpifa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008953.2 | NP_032979.1 | 5 | 534 | ||
GenBank mRNA | AK009032.1 | - | 5 | 546 | ||
AK009654.1 | - | 5 | 546 | |||
AK010181.1 | - | 5 | 546 | |||
BC010288.1 | - | 5 | 534 | 2616413 | ||
BC037124.1 | - | 5 | 555 | |||
BC053336.1 | - | 5 | 534 | 4190594 | ||
X01697.1 | - | 4 | 490 |
