siRNA Details
Gene Symbol: Ptprcap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016933.3 | NP_058629.2 | 2 | 641 | ||
GenBank mRNA | AI323196.1 | - | 2 | 613 | ||
AI627045.1 | - | 1 | 276 | |||
AK088334.1 | - | 2 | 639 | |||
AK165203.1 | - | 2 | 639 | |||
BC013273.1 | - | 2 | 640 | 4208992 | ||
BC100560.1 | - | 2 | 622 | 832557 | ||
D10105.1 | - | 2 | 617 | |||
U03856.1 | - | 1 | 634 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
