siRNA Details
Gene Symbol: Ttc36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138951.1 | NP_620401.1 | 2 | 361 | ||
GenBank mRNA | AK149384.1 | - | 2 | 392 | ||
AK165811.1 | - | 2 | 259 | |||
BC021608.1 | - | 2 | 273 | 5099426 | ||
BC021613.1 | - | 2 | 367 | 5102393 | ||
BC024432.1 | - | 2 | 361 | 4972715 | ||
BC026825.1 | - | 2 | 308 | 4240199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
