siRNA Details
Gene Symbol: Ptrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008986.2 | NP_033012.1 | 2 | 2289 | ||
GenBank mRNA | AK010169.1 | - | 1 | 274 | ||
BC065792.1 | - | 1 | 55 | |||
BC099493.1 | - | 1 | 327 | |||
BC110698.1 | - | 2 | 2267 | 30608521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
