siRNA Details
Gene Symbol: Rdx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104616.1 | NP_001098086.1 | 11 | 1263 | ||
NM_009041.3 | NP_033067.2 | 11 | 1669 | |||
GenBank mRNA | AK146423.1 | - | 11 | 1327 | ||
AK156370.1 | - | 11 | 1355 | |||
AK162204.1 | - | 11 | 1241 | |||
AK168449.1 | - | 11 | 1340 | |||
BC034438.1 | - | 11 | 1495 | |||
X60672.1 | - | 11 | 1282 |
