siRNA Details
Gene Symbol: Gm13247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243138.1 | NP_001230067.1 | 5 | 978 | ||
NM_001243139.1 | NP_001230068.1 | 4 | 863 |
Gene Symbol: Gm13242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103158.2 | NP_001096628.2 | 5 | 977 | ||
GenBank mRNA | BC096593.1 | - | 5 | 989 | 30459933 | |
CD539619.1 | - | 1 | 173 |
Gene Symbol: Rex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177767.1 | NP_001171238.1 | 5 | 977 | ||
GenBank mRNA | AF051348.1 | - | 990 | |||
BC067397.1 | - | 3 | 732 | 30462971 |
Gene Symbol: Zfp600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177545.2 | NP_001171016.1 | 5 | 884 | ||
NM_001177546.2 | NP_001171017.1 | 5 | 977 | |||
GenBank mRNA | BC108352.1 | - | 5 | 884 | 30462028 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
